
Diethyl(3-pyridyl)borane ≥98.0% (by HPLC)
Catalog # TCIAD2661-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Melting point:159.0 to 164.0 deg-C
Specification Test Results
About this item
:
:147.03 g/mol
:GHS07
CAS Number:89878-14-8
Formula:C₉H₁₄BN
MDL Number:MFCD00012348