

3,6-Dioxa-1,8-octanedithiol ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14970-87-7
:
Formula:C₆H₁₄O₂S₂
MDL Number:MFCD00015873
:182.30 g/mol
Boiling Pt:86°C
Flash Pt:132°C
EINECS:239-044-2
UN:2810
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Iodometric back titration):min. 95.0 %
Specification Test Results
Frequently Bought Together