Order Entry
Netherlands
ContactUsLinkComponent
5,6-Dihydro-1,4-dithiin-2,3-dicarboxylic anhydride ≥98.0% (by GC)
Irritant
5,6-Dihydro-1,4-dithiin-2,3-dicarboxylic anhydride ≥98.0% (by GC)
Supplier:  TCI
Certificates
Irritant
5,6-Dihydro-1,4-dithiin-2,3-dicarboxylic anhydride ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Methanolysis Method):
    min. 96.0 %
  • Melting point:
    110.0 to 114.0 deg-C

Specification Test Results

Frequently Bought Together