
5,6-Dichloro-1-ethyl-2-methylbenzimidazole ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD2556-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:108.0 to 113.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:229.10 g/mol
Boiling Pt:195°C
EINECS:221-797-3
:GHS07
CAS Number:3237-62-5
Formula:C₁₀H₁₀Cl₂N₂
MDL Number:MFCD00022843