
(R)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine ≥96.0% (by HPLC)
Catalog # TCIAD2535-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 96.0 area%
- Melting point:141.0 to 145.0 deg-C
- Specific rotation [a]20/D:-330 to -380 deg (C=1.6, EtOH)
Specification Test Results
About this item
:
:441.34 g/mol
:GHS07
CAS Number:74311-54-9
Formula:C₂₆H₂₈FeNP
MDL Number:MFCD00075286