
1,3-Di(2-pyridyl)-1,3-propanedione ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10198-89-7
:
Formula:C₁₃H₁₀N₂O₂
MDL Number:MFCD01321157
:226.24 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:105.0 to 108.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together