
S-(N,N-Dimethylthiocarbamoyl)thioglycolic acid ≥97.0% (by titrimetric analysis)
Catalog # TCIAD2500-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 97.0 %
- Melting point:148.0 to 152.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:179.25 g/mol
:GHS07
CAS Number:4007-01-6
Formula:C₅H₉NO₂S₂
MDL Number:MFCD00021765