
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:87583-89-9
:
Formula:C₈H₁₈N₂
MDL Number:MFCD00671528
:142.25 g/mol
Storage Temperature:Refrigerator
UN:3259
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to lump
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 98.0 ee%
- Melting point:41.0 to 47.0 deg-C
Specification Test Results
Frequently Bought Together