2-(2,2-Diethoxyethyl)-1,3-propanediol ≥95.0% (by GC)
Catalog # TCIAD2437-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 88.0 %
Specification Test Results
About this item
:
:192.26 g/mol
CAS Number:55387-85-4
Formula:C₉H₂₀O₄
MDL Number:MFCD00671524