
4,6-Dimethyl-2-hydroxypyrimidine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:108-79-2
:
Formula:C₆H₈N₂O
MDL Number:MFCD00006072
:124.14 g/mol
EINECS:203-617-5
:GHS07
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:199.0 to 204.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together