Order Entry
Netherlands
ContactUsLinkComponent
5,6-Dichloro-3-pyridinemethanol ≥96.0% (by GC)
5,6-Dichloro-3-pyridinemethanol ≥96.0% (by GC)
Supplier:  TCI
Certificates
5,6-Dichloro-3-pyridinemethanol ≥96.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Melting point:
    76.0 to 80.0 deg-C

Specification Test Results

Frequently Bought Together