16-Doxylstearic acid ≥97.0% (by HPLC, titration analysis) free radical
Supplier: TCI
Certificates
About this item
CAS Number:53034-38-1
:
Formula:C₂₂H₄₂NO₄
MDL Number:MFCD00010066
:384.58 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:55.0 to 60.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together