

Dicyclohexylmethane-4,4'-diisocyanate (mixture of isomers) ≥90.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:5124-30-1
:
Formula:C₁₅H₂₂N₂O₂
MDL Number:MFCD00015405
:262.35 g/mol
Boiling Pt:168°C
Storage Temperature:Refrigerator
EINECS:225-863-2
UN:2206
ADR:6.1,II
:GHS06
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 90.0 % (total of isomer)
Specification Test Results
Frequently Bought Together