5,5-Dimethyl-1-pyrroline-N-oxide ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3317-61-1
:
Formula:C₆H₁₁NO
MDL Number:MFCD00005279
:113.16 g/mol
Merck Index:14,03393
Storage Temperature:Freezer
EINECS:222-011-1
Specification Test Results
- Appearance:White to Gray to Brown powder to lump
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together