2-Dodecyl-p-cresol ≥98.0% (by GC)
Catalog # TCIAD2303-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:46.0 to 49.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:276.46 g/mol
CAS Number:25912-91-8
Formula:C₁₉H₃₂O
MDL Number:MFCD00191638