

N,N-Dimethyldecylamine ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1120-24-7
:
Formula:C₁₂H₂₇N
MDL Number:MFCD00015060
:185.36 g/mol
Boiling Pt:235°C
Flash Pt:92°C
EINECS:214-302-7
UN:2735
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
Specification Test Results
Frequently Bought Together