
6,6'-Dihydroxy-4,4,4',4',7,7'-hexamethyl-2,2'-spirobichroman ≥98.0% (by HPLC)
TCIAD2286-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:203.0 to 210.0 deg-C
:
:368.47 g/mol
:400-270-3
:3077
:9,III
:GHS09
:40278-59-9
:C₂₃H₂₈O₄
:MFCD00191424