Order Entry
Netherlands
ContactUsLinkComponent
2,3-Dibromo-6,7-dicyanonaphthalene ≥98.0% (by HPLC, total nitrogen)
Toxic
2,3-Dibromo-6,7-dicyanonaphthalene ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD2282-1G
Supplier:  TCI
CAS Number:  
Toxic
2,3-Dibromo-6,7-dicyanonaphthalene ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD2282-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Amber powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Solubility in hot DMF:
    almost transparency

Specification Test Results

About this item

 :
 :335.99 g/mol
UN:3439
ADR:6.1,III
 :GHS06
CAS Number:74815-81-9
Formula:C₁₂H₄Br₂N₂
MDL Number:MFCD00191423