Order Entry
Netherlands
ContactUsLinkComponent
Butyl dichloromethyl ether ≥98.0% (by GC)
Flammable
Irritant
Butyl dichloromethyl ether ≥98.0% (by GC)
Catalog # TCIAD2278-10G
Supplier:  TCI
CAS Number:  
Flammable
Irritant
Butyl dichloromethyl ether ≥98.0% (by GC)
Catalog # TCIAD2278-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :157.03 g/mol
Boiling Pt:50°C
UN:1993
ADR:3,II
 :GHS02
 :GHS07
CAS Number:5312-73-2
Formula:C₅H₁₀Cl₂O
MDL Number:MFCD00191420