4-(4-Dimethylaminostyryl)quinoline ≥96.0% (by HPLC, titration analysis)
Catalog # TCIAD2276-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystaline
- Purity (HPLC):min. 96.0 area%
- Purity (Nonaqueous Titration):min. 96.0 %
- Melting point:140.0 to 144.0 deg-C
Specification Test Results
About this item
:
:274.37 g/mol
CAS Number:897-55-2
Formula:C₁₉H₁₈N₂
MDL Number:MFCD00191349