
5,6-Dichloro-2,3-dicyanopyrazine ≥98.0% (by GC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:56413-95-7
:
Formula:C₆Cl₂N₄
MDL Number:MFCD00191419
:198.99 g/mol
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:178.0 to 184.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together