Order Entry
Netherlands
ContactUsLinkComponent
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose ≥98.0% (by GC)
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose ≥98.0% (by GC)
Supplier:  TCI
Certificates
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    74.0 to 77.0 deg-C
  • Specific rotation [a]20/D:
    +35.0 to +38.0 deg (C=1, CHCl3)
  • Solubility in Toluene:
    almost transparency

Specification Test Results

Frequently Bought Together