
4,4'-Dimethoxybenzhydrylamine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:19293-62-0
:
Formula:C₁₅H₁₇NO₂
MDL Number:MFCD00191409
:243.31 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:60.0 to 63.0 deg-C
Specification Test Results
Frequently Bought Together