
6,8-Dimethoxy-4-methylquinoline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:51049-14-0
:
Formula:C₁₂H₁₃NO₂
MDL Number:MFCD00191629
:203.24 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light red to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 % (calcd.on anh.substance)
- Melting point:64.0 to 66.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together