
2-Chloro-5-methyl-1,4-phenylenediamine ≥98.0% (by GC, titration analysis)
TCIAD2179-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:146.0 to 149.0 deg-C
:
:156.61 g/mol
:226-162-4
:GHS07
:5307-03-9
:C₇H₉ClN₂
:MFCD00190684