
1,8-Diaminopyrene ≥97.0% (by HPLC)
Catalog # TCIAD2168-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
About this item
:
:232.29 g/mol
:GHS07
CAS Number:30269-04-6
Formula:C₁₆H₁₂N₂
MDL Number:MFCD00142575