Order Entry
Netherlands
ContactUsLinkComponent
 
 
N,N-Dibutyl-3-aminophenol ≥95.0% (by GC)
 :  TCI
 
 
N,N-Dibutyl-3-aminophenol ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Orange to Brown to Dark purple clear liquid
  • Purity (GC):
    min. 95.0 %

 

Frequently Bought Together