Order Entry
Netherlands
ContactUsLinkComponent
2-(3,4-Dimethoxyphenyl)-N-methylethylamine ≥95.0% (by GC, titration analysis)
Irritant
2-(3,4-Dimethoxyphenyl)-N-methylethylamine ≥95.0% (by GC, titration analysis)
Catalog # TCIAD2104-25G
Supplier:  TCI
CAS Number:  
Irritant
2-(3,4-Dimethoxyphenyl)-N-methylethylamine ≥95.0% (by GC, titration analysis)
Catalog # TCIAD2104-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 95.0 %
  • Purity (Neutralization titration):
    min. 95.0 %

Specification Test Results

About this item

 :
 :195.26 g/mol
Boiling Pt:100°C
EINECS:222-483-9
 :GHS07
CAS Number:3490-06-0
Formula:C₁₁H₁₇NO₂
MDL Number:MFCD00067696