Order Entry
Netherlands
ContactUsLinkComponent
Ethyl iodoacetate for synthesis
Toxic
Corrosive
Ethyl iodoacetate for synthesis
Catalog # 8.20733.1000
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Ethyl iodoacetate for synthesis
Catalog # 8.20733.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Molar Mass:
    214 g/mol
  • Boiling Point:
    179 °C (1013 hPa)
  • Vapor Pressure:
    0.7 hPa (20 °C)
  • Flash Point:
    76 °C
  • Refractive Index:
    1.5045 (20 °C, 589 nm)
  • Density:
    1.81 g/cm3 (20 °C)

Specification Test Results

About this item

 :
Boiling Pt:179 °C (1013 hPa)
Density:1.81 g/cm³ (20 °C)
UN:2927
ADR:6.1,II
 :214 g/mol
EINECS:210-796-3
 :GHS06
 :GHS05
CAS Number:623-48-3
Formula:C₄H₇IO₂
MDL Number:MFCD00001081