

3,4-Dihydro-2-methoxy-4-methyl-2H-pyran (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAD2053-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Orange to Green clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:128.17 g/mol
Boiling Pt:140°C
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:53608-95-0
Formula:C₇H₁₂O₂
MDL Number:MFCD00143062