1,1-Diethylguanidine sulphate ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:77297-00-8
:
Formule:C₅H₁₃N₃
MDL-nummer:MFCD00035597
:328.43 g/mol
EINECS:278-660-6
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Water:almost transparency
Specificatie testresultaten
Frequently Bought Together