2-Hydroxyethyl-2,4-dihydroxybenzoate ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAD2007-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 97.0 %
- Melting point:136.0 to 139.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:198.17 g/mol
Formula:C₉H₁₀O₅
MDL Number:MFCD00059066