5,6-Diamino-2,4-dihydroxypyrimidine sulphate dihydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAD1958-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (with Total Nitrogen):min. 98.0 % (calcd.on anh.substance)
Specification Test Results
About this item
:
:418.34 g/mol
EINECS:264-589-8
CAS Number:63981-35-1
Formula:C₈H₁₂N₈O₄·H₂SO₄·2H₂O
MDL Number:MFCD00135845