
4,5-Dimethyl-1,3-dioxol-2-one ≥99.0% (by GC)
Catalog # TCIAD1920-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light gray to Light yellow powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:78.0 to 81.0 deg-C
Specification Test Results
About this item
:
:114.0993 g/mol
Boiling Pt:130°C
:GHS07
CAS Number:37830-90-3
Formula:C₅H₆O₃
MDL Number:MFCD00135139