
N-(3-Dimethylaminopropyl)methacrylamide ≥98.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:5205-93-6
:
Formula:C₉H₁₈N₂O
MDL Number:MFCD00038359
:170.26 g/mol
Boiling Pt:130°C
Flash Pt:140°C
:Stabilised with 4-methoxyphenol (MEHQ)
EINECS:226-002-3
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Bromination Method):min. 97.0 %
Specification Test Results
Frequently Bought Together