

3-(Dimethylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:69478-75-7
:
Formula:C₆H₁₄N₂
MDL Number:MFCD01569246
Boiling Pt:164°C
UN:2734
ADR:8,II
:114.19 g/mol
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together