Order Entry
Netherlands
ContactUsLinkComponent
1,1'-Di-n-octyl-4,4'-bipyridinium dibromide ≥98.0% (by HPLC, titration analysis)
1,1'-Di-n-octyl-4,4'-bipyridinium dibromide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD1854-5G
Supplier:  TCI
CAS Number:  
1,1'-Di-n-octyl-4,4'-bipyridinium dibromide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD1854-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

 :
 :542.44 g/mol
CAS Number:36437-30-6
Formula:C₂₆H₄₂Br₂N₂
MDL Number:MFCD00060051