Order Entry
Netherlands
ContactUsLinkComponent
2-Butoxyethyl-4-(dimethylamino)benzoate ≥95.0% (by GC)
Irritant
2-Butoxyethyl-4-(dimethylamino)benzoate ≥95.0% (by GC)
Catalog # TCIAD1848-25G
Supplier:  TCI
CAS Number:  
Irritant
2-Butoxyethyl-4-(dimethylamino)benzoate ≥95.0% (by GC)
Catalog # TCIAD1848-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :265.35 g/mol
EINECS:266-668-2
 :GHS07
CAS Number:67362-76-9
Formula:C₁₅H₂₃NO₃
MDL Number:MFCD00055652