2,6-Dimethylpyridinium-p-toluenesulphonate
Catalog # TCIAD1771-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystaline
- Purity (Neutralization titration):min. 98.0 %
- Melting point:115.0 to 119.0 deg-C
- Solubility in Water:almost transparency
- Water:max. 1.0 %
Specification Test Results
About this item
:
:279.35 g/mol
CAS Number:93471-41-1
Formula:C₁₄H₁₇NO₃S
MDL Number:MFCD00060191