
2,7-Dihydroxy-9-fluorenone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:42523-29-5
:
Formula:C₁₃H₈O₃
MDL Number:MFCD00037141
:212.204 g/mol
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together