

Disodium-4,4'-diazidostilbene-2,2'-disulphonate tetrahydrate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAD1606-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 % (calcd.on anh.substance)
- Water:12.0 to 17.0 %
Specification Test Results
About this item
:
:466.35 g/mol
EINECS:220-316-4
UN:1325
ADR:4.1,III
:GHS02
:GHS07
CAS Number:2718-90-3
Formula:C₁₄H₁₀N₆O₆S₂
MDL Number:MFCD02091646