
1,1'-Diheptyl-4,4'-bipyridinium dibromide ≥98.0% (by HPLC, titration analysis) for electrochromic material
Supplier: TCI
Certificates
About this item
CAS Number:6159-05-3
:
Formula:C₂₄H₃₈Br₂N₂
MDL Number:MFCD00011997
:514.39 g/mol
EINECS:228-178-7
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together