Order Entry
Netherlands
ContactUsLinkComponent
 
1,4-Bis(2-hydroxyisopropyl)benzene ≥97.0% (by GC)
 :  TCI
 
1,4-Bis(2-hydroxyisopropyl)benzene ≥97.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    143.0 to 146.0 deg-C

 

Frequently Bought Together