
(R)-2,3-Diaminopropanoic acid hydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:6018-56-0
:
Formula:C₃H₉ClN₂O₂
MDL Number:MFCD00070606
:140.5688 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Specific rotation [a]20/D:-20.0 to -26.0 deg (C=2, 1mol/L HCl)
Specification Test Results
Related Information
Frequently Bought Together