Diveratryl Ether ≥98.0% (by GC)
Catalog # TCIAD1551-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:71.0 to 75.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
About this item
:
:318.37 g/mol
CAS Number:23702-54-7
Formula:C₁₈H₂₂O₅
MDL Number:MFCD00025741