
4,6-Di-tert-butyl-m-cresol ≥96.0% (by GC)
Catalog # TCIAD1432-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 96.0 %
- Melting point:59.0 to 64.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:220.36 g/mol
Merck Index:14,02837
Boiling Pt:282°C
EINECS:207-847-7
:GHS07
Storage Temperature:Refrigerator
CAS Number:497-39-2
Formula:C₁₅H₂₄O
MDL Number:MFCD00059208