
N,N'-Bis(salicylidene)-1,3-propanediamine ≥99.0% (by GC, titration analysis)
TCIAD1422-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
- Melting point:50.0 to 54.0 deg-C
- Solubility in Toluene:almost transparency
:
:282.34 g/mol
:204-418-6
:GHS07
:120-70-7
:C₁₇H₁₈N₂O₂
:MFCD00002245