
4-Dimethylaminobenzenediazosulphonic acid sodium salt ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAD1394-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Ion exchange titration):min. 97.0 % (calcd.on anh.substance)
- Water:max. 8.0 %
Specification Test Results
About this item
:
:251.24 g/mol
EINECS:205-419-4
UN:2811
ADR:6.1,III
:GHS06
CAS Number:140-56-7
Formula:C₈H₁₁N₃O₃S
MDL Number:MFCD00059900