cis-1,4-Diacetoxy-2-butene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:25260-60-0
:
Formula:C₈H₁₂O₄
MDL Number:MFCD00059339
:172.18 g/mol
Boiling Pt:110°C
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together