
Di-p-tolylmethane ≥99.0% (by GC)
Catalog # TCIAD1312-25ML
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
About this item
:
Boiling Pt:150°C
UN:2811
ADR:6.1,II
:196.29 g/mol
:GHS06
CAS Number:4957-14-6
Formula:C₁₅H₁₆
MDL Number:MFCD00026018